Order Entry
Italy
ContactUsLinkComponent
6A-Azido-6A-deoxy-beta-cyclodextrin ≥85.0% (by HPLC)
6A-Azido-6A-deoxy-beta-cyclodextrin ≥85.0% (by HPLC)
Catalog # TCIAA3090-100MG
Supplier:  TCI
CAS Number:  
6A-Azido-6A-deoxy-beta-cyclodextrin ≥85.0% (by HPLC)
Catalog # TCIAA3090-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC) (CAD):
    min. 85.0 area%

Specification Test Results

About this item

 :
 :1160.00 g/mol
Melting Pt:206°C
Storage Temperature:Freezer
CAS Number:98169-85-8
Formula:C₄₂H₆₉N₃O₃₄
MDL Number:MFCD05864973