1-(3-Amino-4-chlorophenyl)ethanone ≥96.0% (by GC, titration analysis)
: TCI
: 1-(3-Amino-4-chlorophenyl)ethan-1-one, 3'-Amino-4'-chloroacetophenone
:79406-57-8
:
:C₈H₈ClNO
:MFCD00122619
:169.61 g/mol
:GHS07
- Environmentally Preferable:
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
- Melting point:105.0 to 109.0 deg-C
Frequently Bought Together