(1R,2R)-2-Aminocyclopentanol hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAA2990-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Specific rotation [a]20/D:-32.0 to -36.0 deg (C=1, H2O)
Specification Test Results
About this item
Synonyms: (1R,2R)-2-aminocyclopentanol hydrochloride, (1|R|,2|R|)-2-Amino-1-hydroxycyclopentane Hydrochloride, trans-(1R,2R)-2-Aminocyclopentanol Hydrochloride, (1R,2R)-2-Amino-1-hydroxycyclopentane hydrochloride
:
:137.61 g/mol
Melting Pt:180°C
:GHS07
Storage Temperature:Refrigerator
CAS Number:68327-11-7
Formula:C₅H₁₁NO·HCl
MDL Number:MFCD09834692