
6-Acetyl-2-Benzoxazolinone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:54903-09-2
:
Formula:C₉H₇NO₃
MDL Number:MFCD01664312
:177.16 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:234.0 to 238.0 deg-C
Specification Test Results
Frequently Bought Together