Order Entry
Italy
ContactUsLinkComponent
4-Anilino-1,1':4',1''-terphenyl ≥98.0% (by HPLC, total nitrogen)
4-Anilino-1,1':4',1''-terphenyl ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAA2903-1G
Supplier:  TCI
CAS Number:  
undefined
4-Anilino-1,1':4',1''-terphenyl ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAA2903-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    193.0 to 197.0 deg-C

Specification Test Results

About this item

 :
 :321.42 g/mol
CAS Number:897671-81-7
Formula:C₂₄H₁₉N
MDL Number:MFCD28963769