Afatinib ≥97.0% (by HPLC, total nitrogen)
: TCI
: (S,E)-N-(4-((3-Chloro-4-fluorophenyl)amino)-7-((tetrahydrofuran-3-yl)oxy)quinazolin-6-yl)-4-(dimethylamino)but-2-enamide, BIBW2992, Tovok
:850140-72-6
:C₂₄H₂₅ClFN₅O₃
:MFCD12407405
:485.94 g/mol
:102°C
:Refrigerator
- Environmentally Preferable:
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Water:max. 3.0 %
Frequently Bought Together