

Palmitoyl chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-67-4
:
Formula:C₁₆H₃₁ClO
MDL Number:MFCD00000742
:274.87 g/mol
Boiling Pt:324 °C (1013 hPa)
Melting Pt:12 °C
Density:0.91 g/cm³ (20 °C)
Flash Pt:182 °C
UN:3265
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:203-996-7
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:12 °C
- Molar Mass:274.87 g/mol
- Boiling Point:174 °C (5.1 hPa)
- Flash Point:182 °C
- Density:0.91 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together