Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
11-Azidoundecyltrimethoxysilane
11-Azidoundecyltrimethoxysilane ≥93.0% (by GC)
11-Azidoundecyltrimethoxysilane ≥93.0% (by GC)
TCIAA2783-100MG
:
TCI
:
11-Azidoundecyltrimethoxysilane ≥93.0% (by GC)
TCIAA2783-100MG
:
TCI
:
Pk:
100 mg
Seq:
2440
Appearance:
Colorless to Light yellow clear liquid
Purity (GC):
min. 93.0 %
:
:
317.505 g/mol
:
GHS07
:
334521-23-2
:
C₁₄H₃₁N₃O₃Si
:
MFCD11982878