Order Entry
Italy
ContactUsLinkComponent
Azulene-1-carboxaldehyde ≥97.0% (by GC)
Azulene-1-carboxaldehyde ≥97.0% (by GC)
Catalog # TCIAA2752-1G
Supplier:  TCI
CAS Number:  
Azulene-1-carboxaldehyde ≥97.0% (by GC)
Catalog # TCIAA2752-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Dark green to Dark purple to Black clear liquid to cloudy liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :156.18 g/mol
CAS Number:7206-61-3
Formula:C₁₁H₈O
MDL Number:MFCD19301077