
2-Amino-4-methoxypyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10201-73-7
:
Formula:C₆H₈N₂O
MDL Number:MFCD07437849
:124.14 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:116.0 to 120.0 deg-C
Specification Test Results
Frequently Bought Together