Order Entry
Italy
ContactUsLinkComponent
4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid hydrate ≥98.0% (by HPLC, titration analysis)
Irritant
4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid hydrate ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic acid hydrate ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Gray to Dark blue powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    4.0 to 8.0 %

Specification Test Results

Frequently Bought Together