Methyl-3-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
: TCI
:3196-73-4
:
:C₄H₉NO₂·HCl
:MFCD00039060
:139.6 g/mol
:Refrigerator
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Frequently Bought Together