

2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane ≥98.0% (by GC, titration analysis) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:911482-75-2
:
Formula:C₈H₁₅BO₂
:154.02 g/mol
ADR:3,III
:Stabilised with phenothiazine
UN:1993
:GHS02
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together