
5-Amino-2-methylbenzenesulphonamide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6973-09-7
:
Formula:C₇H₁₀N₂O₂S
MDL Number:MFCD06681071
:186.23 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:165.0 to 170.0 deg-C
Specification Test Results
Frequently Bought Together