
3-Amino-3-(3'-pyridyl)propionic acid ≥98.0% (by GC, titration analysis)
TCIAA2537-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:205.0 to 209.0 deg-C
:
:166.1800 g/mol
:GHS07
:62247-21-6
:C8H10N2O2
:MFCD01549598