Order Entry
Italy
ContactUsLinkComponent
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol ≥98.0% (by HPLC, titration analysis)
Irritant
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA2533-1G
Supplier:  TCI
CAS Number:  
Irritant
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA2533-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Optical purity (LC):
    min. 99.0 ee%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    96.0 to 101.0 deg-C

Specification Test Results

About this item

 :
 :255.3600 g/mol
 :GHS07
CAS Number:86695-06-9
Formula:C₁₇H₂₁NO
MDL Number:MFCD01318247