Order Entry
Italy
ContactUsLinkComponent
 
Irritant
5-Amino-α,α,α-trifluoro-m-toluic acid ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAA2481-1G
Supplier:  TCI
CAS Number:  
Irritant
5-Amino-α,α,α-trifluoro-m-toluic acid ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAA2481-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    141.0 to 145.0 deg-C

Specification Test Results

About this item

 :
 :205.14 g/mol
 :GHS07
CAS Number:328-68-7
Formula:C₈H₆F₃NO₂
MDL Number:MFCD00236641