2-Azido-1,3-bis[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]propane ≥95.0% (by HPLC)
TCIAA2474-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White or Colorless to Light orange to Yellow powder to lump to clear liquid
- Purity (HPLC) (CAD):min. 95.0 area%
:
:345.40 g/mol
:Refrigerator
:1392500-07-0
:C₁₅H₂₇N₃O₆