
Amyltriethylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Catalog # TCIAA2274-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Yellow clear liquid
- Purity (Ion exchange titration):min. 98.0 %
Specification Test Results
About this item
:
:452.47 g/mol
:GHS07
CAS Number:906478-91-9
Formula:C₁₃H₂₆F₆N₂O₄S₂
MDL Number:MFCD15072143