O-Acetyl-N-carbobenzoxyhydroxylamine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:180798-01-0
:
Formula:C₁₀H₁₁NO₄
:209.20 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together