Order Entry
Italy
ContactUsLinkComponent
Nɑ-Acetyl-L-lysine ≥97.0% (by HPLC, titration analysis)
Nɑ-Acetyl-L-lysine ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Nɑ-Acetyl-L-lysine ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Specific rotation [a]20/D:
    +3.0 to +5.0 deg (C=4, H2O)

Specification Test Results

Related Information

Frequently Bought Together