Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Aminoadenosine ≥98.0% (by HPLC, titration analysis)

2-Aminoadenosine ≥98.0% (by HPLC, titration analysis)

Irritant
Nome del fornitore:   TCI
2-Aminoadenosine ≥98.0% (by HPLC, titration analysis)
Nome del fornitore:  TCI
Certificates
2-Aminoadenosine ≥98.0% (by HPLC, titration analysis)

2 Options Available Below

Product Details & Documents
Synonyms:  2,6-Diamino-9-(beta-D-ribofuranosyl)purine, (2R,3R,4S,5R)-2-(2,6-Diamino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 2,6-Diaminopurine Riboside
  • CAS Number:2096-10-8
  • Formula:C₁₀H₁₄N₆O₄
  • MDL Number:MFCD00053556
  • Molecular Weight:282.26 g/mol
  • Melting Pt:252°C
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -36.0 to -44.0 deg (C=1, 0.1mol/L HCl)
Product Family Options

Product Information

  • Pk
    Disponibilità
    Price
  • 5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 58,00 €
    58,00 €
    1 * 5 g
       58,00 €

  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 225,00 €
    225,00 €
    1 * 25 g
       225,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.