
1-Allyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:65039-10-3
:
Formula:C₇H₁₁ClN₂
MDL Number:MFCD07784450
:158.63 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Brown clear liquid
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together