Order Entry
Italy
ContactUsLinkComponent
(R)-1-Acetyl-3-pyrrolidinol ≥98.0% (by GC)
Irritant
(R)-1-Acetyl-3-pyrrolidinol ≥98.0% (by GC)
Catalog # TCIAA2049-1G
Supplier:  TCI
CAS Number:  
Irritant
(R)-1-Acetyl-3-pyrrolidinol ≥98.0% (by GC)
Catalog # TCIAA2049-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    81.0 to 85.0 deg-C
  • Specific rotation [a]20/D:
    -39.0 to -45.0 deg (C=3, MeOH)

Specification Test Results

About this item

 :
 :129.16 g/mol
 :GHS07
CAS Number:916733-17-0
Formula:C₆H₁₁NO₂
MDL Number:MFCD09909313