Order Entry
Italy
ContactUsLinkComponent
Ammonia ∼4% ∼2.0 mol/L in methanol
Flammable
Irritant
Health
Environment
Ammonia ∼4% ∼2.0 mol/L in methanol
Catalog # TCIAA1884-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
Ammonia ∼4% ∼2.0 mol/L in methanol
Catalog # TCIAA1884-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Concentration (Neutralization titration):
    4.0 to 4.5 w/w%

Specification Test Results

About this item

 :
 :17.03 g/mol
Merck Index:14,00492
Flash Pt:6°C
Storage Temperature:Refrigerator
EINECS:231-635-3
UN:1992
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:7664-41-7
Formula:NH₃
MDL Number:MFCD00011418