

Acid red 114 ≥80.0% (by HPLC)
Catalog # TCIAA1874-25G
Supplier: TCI
CAS Number:
Specifications
- Conf:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 80.0 area%
- Lambda max.:513.0 to 517.0 nm (EtOH (50%))
- Absorbance (E1%1cm):min. 300 (EtOH (50%), 513.0 to 517.0nm)
Specification Test Results
About this item
:
:830.81 g/mol
EINECS:229-272-0
UN:3077
ADR:9,III
:GHS08
:GHS09
CAS Number:6459-94-5
Formula:C₃₇H₃₀N₄O₁₀S₃
MDL Number:MFCD00003894