Order Entry
Italy
ContactUsLinkComponent
 
1,4-Anhydroerythritol ≥98.0% (by GC)
  TCIAA1839-5G
 :  TCI
 :  
1,4-Anhydroerythritol ≥98.0% (by GC)
  TCIAA1839-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :104.11 g/mol
 :224-433-1
 :4358-64-9
 :C₄H₈O₃
 :MFCD00075210