Order Entry
Italy
ContactUsLinkComponent
 
4-(3-Aminopropyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
 :  TCI
 
4-(3-Aminopropyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

Frequently Bought Together