4-Aminothiomorpholine-1,1-dioxide ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:26494-76-8
:
Formula:C₄H₁₀N₂O₂S
:150.20 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:105.0 to 110.0 deg-C
Specification Test Results
Frequently Bought Together