Order Entry
Italy
ContactUsLinkComponent
2-Acetamido-3,4,6-tri-O-benzyl-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)
2-Acetamido-3,4,6-tri-O-benzyl-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
2-Acetamido-3,4,6-tri-O-benzyl-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Specific rotation [a]20/D:
    +4.0 to +8.0 deg (C=1, CHCl3)

Specification Test Results

Frequently Bought Together