Order Entry
Italy
ContactUsLinkComponent
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0% (by GC, titration analysis)
Irritant
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    117.0 to 121.0 deg-C
  • Specific rotation [a]20/D:
    -41.0 to -45.0 deg (C=1, MeOH)

Specification Test Results

Related Information

Frequently Bought Together