
Ethyl-4-amino-1-piperidinecarboxylate ≥97.0% (by GC)
: TCI
:58859-46-4
:
:C₈H₁₆N₂O₂
:MFCD00006484
:172.23 g/mol
:78°C
:261-472-3
:GHS07
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together