6-Acetyl-1-bromo-2-methoxynaphthalene ≥98.0% (by GC)
Catalog # TCIAA1584-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
About this item
:
:279.13 g/mol
CAS Number:84167-74-8
Formula:C₁₃H₁₁BrO₂
MDL Number:MFCD00100832