
Azobenzene-4,4'-dicarbonyl dichloride ≥98.0% (by titrimetric analysis)
: TCI
:10252-29-6
:
:C₁₄H₈Cl₂N₂O₂
:MFCD00053223
:307.13 g/mol
:3261
:8,III
:GHS05
- Appearance:Dark red to Brown powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:162.0 to 166.0 deg-C
Frequently Bought Together