N-Acetyl-L-phenylalanine ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2018-61-3
:
Formula:C₁₁H₁₃NO₃
MDL Number:MFCD00063158
:207.2 g/mol
EINECS:217-959-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:163.0 to 167.0 deg-C
- Specific rotation [a]20/D:+38.0 to +42.0 deg (C=5, MeOH)
Specification Test Results
Frequently Bought Together