Order Entry
Italy
ContactUsLinkComponent
 
 
1,3-Adamantanediol ≥98.0% (by GC)
 :  TCI
 
 
1,3-Adamantanediol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

 

 

Frequently Bought Together