Order Entry
Italy
ContactUsLinkComponent
(3-Acrylamidopropyl)trimethylammonium chloride 74-76% in water stabilised
Irritant
(3-Acrylamidopropyl)trimethylammonium chloride 74-76% in water stabilised
Supplier:  TCI
Certificates
undefined
Irritant
(3-Acrylamidopropyl)trimethylammonium chloride 74-76% in water stabilised
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Brown clear liquid
  • Concentration (Bromination Method):
    74.0 to 76.0 w/w%
  • Water:
    24.0 to 26.0 %

Specification Test Results

Related Information

Frequently Bought Together