S-Allyl-L-cysteine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:21593-77-1
:
Formula:C₆H₁₁NO₂S
MDL Number:MFCD00151975
:161.22 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 95.0 area%
- Optical purity (LC):min. 99.0 ee%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together