
N-Acetyl-L-tyrosine ≥99.0% (by titrimetric analysis)
Catalog # TCIAA1409-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:151.0 to 154.0 deg-C
- Specific rotation [a]20/D:+44.0 to +50.0 deg (C=2, H2O)
Specification Test Results
About this item
:
:223.23 g/mol
EINECS:208-671-3
:GHS05
CAS Number:537-55-3
Formula:C₁₁H₁₃NO₄
MDL Number:MFCD00037190