Order Entry
Italy
ContactUsLinkComponent
Acid violet 43 ≥97.0% (by HPLC)
Irritant
Acid violet 43 ≥97.0% (by HPLC)
Catalog # TCIAA1402-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Acid violet 43 ≥97.0% (by HPLC)
Catalog # TCIAA1402-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Dark purple powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Lambda max.:
    565.0 to 571.0 nm (H2O)
  • Absorbance (E1%1cm):
    min. 160 (H2O, 565.0 to 571.0 nm)

Specification Test Results

About this item

 :
 :431.39 g/mol
EINECS:224-618-7
 :GHS07
CAS Number:4430-18-6
Formula:C₂₁H₁₅NO₆S
MDL Number:MFCD00068446