Order Entry
Italy
ContactUsLinkComponent
β-Alaninamide hydrochloride ≥98.0% (by total nitrogen and titration analysis)
β-Alaninamide hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier:  TCI
Certificates
β-Alaninamide hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    149.0 to 154.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together