Di-n-alkyl adipate plasticizer
Catalog # TCIAA1386-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
Specification Test Results
About this item
:
MDL Number:MFCD00675047