
N-(tert-Butoxycarbonyl)-1,4-diaminobutane ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:68076-36-8
:
Formula:C₉H₂₀N₂O₂
MDL Number:MFCD00210019
:188.27 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together