Order Entry
Italy
ContactUsLinkComponent
 
 
(S)-(-)-2-Acetoxypropionic acid ≥98.0% (by titrimetric analysis)
 :  TCI
 
 
(S)-(-)-2-Acetoxypropionic acid ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 94.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -44.0 to -55.0 deg (C=7, CHCl3)

 

Frequently Bought Together