6-Amidino-2-naphthol methanesulphonate ≥99.0% (by HPLC, titration analysis)
: TCI
:82957-06-0
:
:C₁₁H₁₀N₂O
:MFCD00143493
:282.31 g/mol
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Solubility in Methanol:almost transparency
Frequently Bought Together