
S-(2-Aminoethyl)isothiouronium bromide hydrobromide ≥98.0% (by titrimetric analysis)
Catalog # TCIAA1078-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:281.01 g/mol
Merck Index:14,00178
EINECS:200-257-0
:GHS07
CAS Number:56-10-0
Formula:C₃H₁₁Br₂N₃S
MDL Number:MFCD00037011