Order Entry
Italy
ContactUsLinkComponent
 
Flammable
Corrosive
Irritant
Health
Allylmagnesium bromide ∼13% ∼0.7 mol/L in ethyl ether
Catalog # TCIAA0963-100ML
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
Irritant
Health
Allylmagnesium bromide ∼13% ∼0.7 mol/L in ethyl ether
Catalog # TCIAA0963-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Concentration (sec Butanol method):
    12.5 to 14.0 w/w%

Specification Test Results

About this item

 :
 :145.28 g/mol
EINECS:217-046-4
UN:3399
ADR:4.3,I
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:1730-25-2
Formula:C₃H₅BrMg
MDL Number:MFCD00000044