Order Entry
Italy
ContactUsLinkComponent
Dipropyl adipate ≥98.0% (by GC)
Dipropyl adipate ≥98.0% (by GC)
Catalog # TCIAA0931-25ML
Supplier:  TCI
CAS Number:  
Dipropyl adipate ≥98.0% (by GC)
Catalog # TCIAA0931-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :230.30 g/mol
Boiling Pt:155°C
Flash Pt:146°C
EINECS:203-371-9
CAS Number:106-19-4
Formula:C₁₂H₂₂O₄
MDL Number:MFCD00048696