Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Diallyl adipate

25 mL
  TCIAA0824-25ML
 
 :   TCI
Diallyl adipate
  TCIAA0824-25ML
 :  TCI
 :  2998-04-1
Diallyl adipate

25 mL

 :  2998-04-1
 
56,00 €
   
1 * 25 mL
 
 :  Adipic Acid Diallyl Ester
  •  :2998-04-1
  •  :C₁₂H₁₈O₄
  •  :MFCD00039826
  •  :226.27 g/mol
  •  :149°C
  •  :221-071-6
 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

  • Pk:
    25 mL