Order Entry
Italy
ContactUsLinkComponent
Aluminium tri-tert-butanolate ≥98.0% (by titrimetric analysis)
 
 
Aluminium tri-tert-butanolate ≥98.0% (by titrimetric analysis)
  TCIAA0804-5G
 :  TCI
 :  
 
 
Aluminium tri-tert-butanolate ≥98.0% (by titrimetric analysis)
  TCIAA0804-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Gray powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %

 

 

 :
 :246.33 g/mol
 :14,00333
 :209-146-1
 :2925
 :4.1,III
 :GHS02
 :GHS05
 :556-91-2
 :C₁₂H₂₇AlO₃
 :MFCD00008801